Learn More
Cayman Chemical Z-YVAD-CMK (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502937704
Encompass_Preferred
An inhibitor of caspase-1 and caspase-3; reduces IL-1β and IL-18 secretion in isolated mouse peritoneal macrophages by cholesterol-treated rPLY at 30 µM; reduces caspase-3 activation by the EGF-like domain of tenascin-C in detached hSMCs in vitro; decreases cell death induced by the EGF-like domain of tenascin-C by 50% at 100 µM; completely inhibits cell death of hSMCs at 200-400 µM
Catalog No. 50-293-7704 Supplier Cayman Chemical Supplier No. 274351 MG
An inhibitor of caspase-1 and caspase-3; reduces IL-1β and IL-18 secretion in isolated mouse peritoneal macrophages by cholesterol-treated rPLY at 30 µM; reduces caspase-3 activation by the EGF-like domain of tenascin-C in detached hSMCs in vitro; decreases cell death induced by the EGF-like domain of tenascin-C by 50% at 100 µM; completely inhibits cell death of hSMCs at 200-400 µM