Learn More
Apexbio Technology LLC Z-YVAD-FMK 210344-97-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061526
Encompass_Preferred
Z-YVAD-FMK (CAS 210344-97-1) is a cell-permeable irreversible inhibitor of caspase-1 By inhibiting caspase-1 activity it interferes with caspase-mediated signaling cascades involved in cell apoptosis and cytokine maturation In Caco-2 cells Z-YVAD-FMK reduces butyrate-induced growth inhibition and fully abrogates butyrate s effects at 100 mol/L Additionally in MyD88-deficient murine bone marrow dendritic cells Z-YVAD-FMK effectively blocks alum-induced caspase-1 activation and subsequent IL-1 release It serves as a valuable research tool for investigating caspase-dependent apoptotic pathways and inflammatory responses
Catalog No. 50-000-61526 Supplier Apexbio Technology LLC Supplier No. A89555.1
Z-YVAD-FMK (CAS 210344-97-1) is a cell-permeable irreversible inhibitor of caspase-1 By inhibiting caspase-1 activity it interferes with caspase-mediated signaling cascades involved in cell apoptosis and cytokine maturation In Caco-2 cells Z-YVAD-FMK reduces butyrate-induced growth inhibition and fully abrogates butyrate s effects at 100 mol/L Additionally in MyD88-deficient murine bone marrow dendritic cells Z-YVAD-FMK effectively blocks alum-induced caspase-1 activation and subsequent IL-1 release It serves as a valuable research tool for investigating caspase-dependent apoptotic pathways and inflammatory responses