Learn More
Medchemexpress LLC Z-YVAD-FMK | 210344-97-1 | 98.0% | 630.66 | 10 MG
SDP
Catalog No. 5000424644
Encompass_Preferred
Z-YVAD-FMK is a cell-permeable caspase-1 and -4 inhibitor, demonstrating both anti-inflammatory and anti-tumor activities. It is intended for research use only and is not suitable for medical applications or sale to patients.
  • Cell-permeable caspase-1 and -4 inhibitor
  • Exhibits anti-inflammatory properties
  • Demonstrates anti-tumor activities
  • Suitable for various research applications
Catalog No. 50-004-24644 Supplier Medchemexpress LLC Supplier No. HYP100910MG
Z-YVAD-FMK is a cell-permeable caspase-1 and -4 inhibitor, demonstrating both anti-inflammatory and anti-tumor activities. It is intended for research use only and is not suitable for medical applications or sale to patients.
  • Cell-permeable caspase-1 and -4 inhibitor
  • Exhibits anti-inflammatory properties
  • Demonstrates anti-tumor activities
  • Suitable for various research applications