Learn More
Enzo Life Sciences Z-YVAD-FMK (Ready-to-Use) (100µl)
SDP
Catalog No. 502008359
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4. Alternative name: Caspase 1 inhibitor (fluoromethylketone)or. MW: 630.7. Purity: ≥99% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8359 Supplier Enzo Life Sciences Supplier No. ALX260154R100
Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4. Alternative name: Caspase 1 inhibitor (fluoromethylketone)or. MW: 630.7. Purity: ≥99% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.