Learn More
Medchemexpress LLC Z,Z-Dienestrol-d6 | 91297-99-3 | 99.8% | 272.37 | 1 MG
SDP
Catalog No. 5000349635
Encompass_Preferred
Z,Z-Dienestrol-d6 is the deuterium labeled Z,Z-Dienestrol. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-49635 Supplier Medchemexpress LLC Supplier No. HY139507S1MG
Add to Cart
Add to Cart
Z,Z-Dienestrol-d6 is the deuterium labeled Z,Z-Dienestrol. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.