Learn More
Medchemexpress LLC (Z)-Zolucatetide ((Z)-FOG-001; I-67) | 98.2% | 2076.39 | 5 MG
SDP
Catalog No. 5000424073
Encompass_Preferred
(Z)-Zolucatetide is a structural isomer of Zolucatetide and acts as a potent β-catenin inhibitor with an IC50 of <50 nM. It inhibits β-catenin and T-cell factor (TCF) transcription factor interaction, suppresses cell proliferation, and induces cell cycle arrest in target cells. It has shown anti-tumor efficacy in mice bearing COLO320DM colon cancer cell xenografts and can be used for the study of colon cancer.
  • Potent β-catenin inhibitor with an IC50 of less than 50 nM.
  • Inhibits β-catenin and T-cell factor (TCF) transcription factor interaction.
  • Suppresses cell proliferation.
  • Induces cell cycle arrest in target cells.
  • Exhibits anti-tumor efficacy in mice bearing COLO320DM colon cancer cell xenografts.
  • Useful for the study of colon cancer.
Catalog No. 50-004-24073 Supplier Medchemexpress LLC Supplier No. HYP10925A5MG
Add to Cart
Add to Cart
(Z)-Zolucatetide is a structural isomer of Zolucatetide and acts as a potent β-catenin inhibitor with an IC50 of <50 nM. It inhibits β-catenin and T-cell factor (TCF) transcription factor interaction, suppresses cell proliferation, and induces cell cycle arrest in target cells. It has shown anti-tumor efficacy in mice bearing COLO320DM colon cancer cell xenografts and can be used for the study of colon cancer.
  • Potent β-catenin inhibitor with an IC50 of less than 50 nM.
  • Inhibits β-catenin and T-cell factor (TCF) transcription factor interaction.
  • Suppresses cell proliferation.
  • Induces cell cycle arrest in target cells.
  • Exhibits anti-tumor efficacy in mice bearing COLO320DM colon cancer cell xenografts.
  • Useful for the study of colon cancer.