Learn More
Montana Molecular Human Somatostatin Receptor Type 2 (SSTR2) Big Sky BacMam, 15mL
SDP
Catalog No. 502815268
Encompass_Preferred
Human Somatostatin Receptor Type 2 packaged in Big Sky BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5268 Supplier Montana Molecular Supplier No. Z2109N015
Human Somatostatin Receptor Type 2 packaged in Big Sky BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.