Learn More
Montana Molecular Human M2 Muscarinic Receptor (hM2R, CHRM2, ACM2) Big Sky BacMam, 15mL
SDP
Catalog No. 502815258
Encompass_Preferred
Human M2 muscarinic acetylcholine packaged in Big Sky BacMam for mammalian cell expression. Big Sky BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5258 Supplier Montana Molecular Supplier No. Z0219N015
Human M2 muscarinic acetylcholine packaged in Big Sky BacMam for mammalian cell expression. Big Sky BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.