Learn More
Montana Molecular Human Adenosine A1 Receptor (AA1R, ADORA1) Big Sky BacMam, 15mL
SDP
Catalog No. 502815271
Encompass_Preferred
Human A1 Adenosine receptor packaged in Montana Molecular's Big Sky BacMam vector. Express the adenosine receptor in your cells of interest. Big Sky BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5271 Supplier Montana Molecular Supplier No. Z0309N015
Human A1 Adenosine receptor packaged in Montana Molecular's Big Sky BacMam vector. Express the adenosine receptor in your cells of interest. Big Sky BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.