Learn More
Montana Molecular Human Succinate Receptor (SUCNR1, SUCR1) BacMam, 30mL
SDP
Catalog No. 502815272
Encompass_Preferred
Use this assay to express the human Succinate receptor in your cells of interest. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. Importantly, BacMam expression is easily titrateable to suit your experimental needs. This tool can be co-expressed with Montana Molecular's assays for cAMP to investigate signaling dynamics at the SUCNR1 receptor.
Catalog No. 50-281-5272 Supplier Montana Molecular Supplier No. Z0400N030
Use this assay to express the human Succinate receptor in your cells of interest. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. Importantly, BacMam expression is easily titrateable to suit your experimental needs. This tool can be co-expressed with Montana Molecular's assays for cAMP to investigate signaling dynamics at the SUCNR1 receptor.