Learn More
Montana Molecular Human GLP-1 Receptor (GLP1R) BacMam, 30mL
SDP
Catalog No. 502815240
Encompass_Preferred
Human GLP-1 Receptor packaged in Big Sky BacMam for enhanced expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5240 Supplier Montana Molecular Supplier No. Z0609N015
Human GLP-1 Receptor packaged in Big Sky BacMam for enhanced expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.