Learn More
Montana Molecular Human Kappa Opioid Receptor (KOR, OPRK, OPRK1) Big Sky BacMam, 15mL
SDP
Catalog No. 502815277
Encompass_Preferred
Human Kappa Opioid receptor packaged in Big Sky BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5277 Supplier Montana Molecular Supplier No. Z0719N015
Human Kappa Opioid receptor packaged in Big Sky BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.