Learn More
Montana Molecular Human Cannabinoid Receptor 1 (CB1R, CNR1) Big Sky BacMam, 15mL
SDP
Catalog No. 502815252
Encompass_Preferred
Human Cannabinoid receptor 1 packaged in BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5252 Supplier Montana Molecular Supplier No. Z0739N015
Human Cannabinoid receptor 1 packaged in BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.