Learn More
Montana Molecular Human Delta Opioid Receptor (DOR, OPRD, OPRD1) Big Sky BacMam, 15mL
SDP
Catalog No. 502815253
Encompass_Preferred
Human Delta Opioid receptor packaged in BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5253 Supplier Montana Molecular Supplier No. Z0749N015
Human Delta Opioid receptor packaged in BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.