Learn More
Montana Molecular Human C-C Chemokine Receptor Type 5 (CCR5) BacMam, 30mL
SDP
Catalog No. 502815263
Encompass_Preferred
Use this assay to express the human C-C Chemokine receptor 5 (CCR5) in your cells of interest. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5263 Supplier Montana Molecular Supplier No. Z1010N030
Use this assay to express the human C-C Chemokine receptor 5 (CCR5) in your cells of interest. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.