Learn More
Montana Molecular Human Serotonin 5-HT2A Receptor (HTR2A, 5HT2AR) BacMam, 30mL
SDP
Catalog No. 502815264
Encompass_Preferred
Use this assay to express the human 5-hydroxytryptamine receptor 2A in your cells of interest. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. Importantly, BacMam expression is easily titrateable to suit your experimental needs. This tool can be co-expressed with Montana Molecular's assays for DAG, Calcium, or arrestin to investigate signaling dynamics or receptor activation at the 5-HT2A receptor.
Catalog No. 50-281-5264 Supplier Montana Molecular Supplier No. Z1100N030
Use this assay to express the human 5-hydroxytryptamine receptor 2A in your cells of interest. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. Importantly, BacMam expression is easily titrateable to suit your experimental needs. This tool can be co-expressed with Montana Molecular's assays for DAG, Calcium, or arrestin to investigate signaling dynamics or receptor activation at the 5-HT2A receptor.