Learn More
ATCC Z-138; Mantle Cell Lymphoma; Human (Homo sapiens)
Supplier:  ATCC CRL3001
Encompass
Z-138 is a lymphoblast cell line isolated in 1990 from a White male who initially was diagnosed with chronic lymphocytic leukemia. Approximately 2 years later, the neoplasm had transformed into an aggressive mantle cell lymphoma. It has applications for immunology research.
Catalog No. 50-238-3171