Learn More
Montana Molecular Human Parathyroid Hormone 2 Receptor (PTH2R) Big Sky BacMam, 15mL
SDP
Catalog No. 502815278
Encompass_Preferred
Human parathyroid hormone 2 receptor (PTH2R) packaged in Big Sky BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5278 Supplier Montana Molecular Supplier No. Z1419N015
Human parathyroid hormone 2 receptor (PTH2R) packaged in Big Sky BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.