Learn More
Montana Molecular Human Serotonin 5-HT1B Receptor (HTR1B, 5HT1BR) Big Sky BacMam, 15mL
SDP
Catalog No. 502815233
Encompass_Preferred
Human 5-HT1B packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5233 Supplier Montana Molecular Supplier No. Z1909N015
Human 5-HT1B packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.