Learn More
Alkali Scientific Alkali Scientific™ Z207-A Sprint™ 8 Plus Clinical Centrifuge with 8 x 15ml Fixed Angle Rotor, 115V
SDP
Catalog No. 503029529
Encompass_Preferred
The Alkali Scientific™ Z207-A, unlike traditional clinical centrifuge machines, has an increased speed capability (4445xg). This allows the Sprint™ 8 PLUS to separate serum or plasma in as little as 2-3 minutes. Additional features include: – A large back-lit, liquid crystal display– High capacity rotor, 8 tubes up to 15ml (conical or round bottom)– Unique EZ-Scroll™ touch pad– Timer up to 99hr 99mins. / continuous– Storage of commonly used run-times– Control of speed in in both rpm and rcf– Adjustable acceleration and deceleration levels– Safety imbalance detection system– Momentary operation– Electrical lid lock Ideal for rapid production of serum and plasma
Catalog No. 50-302-9529 Supplier Alkali Scientific Supplier No. RS7536
The Alkali Scientific™ Z207-A, unlike traditional clinical centrifuge machines, has an increased speed capability (4445xg). This allows the Sprint™ 8 PLUS to separate serum or plasma in as little as 2-3 minutes. Additional features include: – A large back-lit, liquid crystal display– High capacity rotor, 8 tubes up to 15ml (conical or round bottom)– Unique EZ-Scroll™ touch pad– Timer up to 99hr 99mins. / continuous– Storage of commonly used run-times– Control of speed in in both rpm and rcf– Adjustable acceleration and deceleration levels– Safety imbalance detection system– Momentary operation– Electrical lid lock Ideal for rapid production of serum and plasma