Learn More
Montana Molecular Human Melanocortin 3 Receptor (MC3R) Big Sky BacMam, 15mL
SDP
Supplier:  Montana Molecular Z2229N015
Encompass_Preferred
Human Melanocortin 3 Receptor (MC3R) packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5265