Learn More
Montana Molecular Human Melanocortin 4 Receptor (MC4R) Big Sky BacMam, 15mL
SDP
Catalog No. 502815246
Encompass_Preferred
Human Melanocortin 4 Receptor (MC4R) packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5246 Supplier Montana Molecular Supplier No. Z2239N015
Human Melanocortin 4 Receptor (MC4R) packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.