Learn More
Montana Molecular Human Apelin Receptor (APLNR) Big Sky BacMam, 15mL
SDP
Catalog No. 502815231
Encompass_Preferred
Human Apelin Receptor (APLNR) packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5231 Supplier Montana Molecular Supplier No. Z2309N015
Human Apelin Receptor (APLNR) packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.