Learn More
Montana Molecular Human ACKR3 Big Sky BacMam, 15mL
SDP
Catalog No. 502815232
Encompass_Preferred
Human Atypical Chemokine Receptor 3 (ACKR3) packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5232 Supplier Montana Molecular Supplier No. Z2509N015
Human Atypical Chemokine Receptor 3 (ACKR3) packaged in Big Sky BacMam for robust expression in mammalian cells. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.