Learn More
Montana Molecular Human CALCRL Big Sky BacMam, 15mL
SDP
Supplier:  Montana Molecular Z2609N015
Encompass_Preferred
Human Calcitonin Receptor-like (CALCRL), also called Calcitonin Receptor-like Receptor (CRLR). Big Sky BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5267