Learn More
Cayman Chemical (Z)-4-hydroxy Tamoxifen, 68047-06-3, 25 mg
SDP
Supplier:  Cayman Chemical 1485425
Encompass_Preferred
A major phase I metabolite of tamoxifen, a well-known estrogen receptor antagonist in breast but partial estrogen receptor agonist in endometrium; has at least a 100-fold higher affinity for estrogen receptors than tamoxifen
Catalog No. NC1355029
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.