Learn More
Cayman Chemical (Z)-4-hydroxy Tamoxifen, 68047-06-3, 5 mg
SDP
Supplier:  Cayman Chemical 148545
Encompass_Preferred
A major phase I metabolite of tamoxifen, a well-known estrogen receptor antagonist in breast but partial estrogen receptor agonist in endometrium; has at least a 100-fold higher affinity for estrogen receptors than tamoxifen
Catalog No. NC1355027