Learn More
Med Vet International Z4 Zoom Stereoscope, Trinocular, Dual LED Stand, 110V, J0755Z4T, Each
Catalog No. 5000584917
Encompass_Preferred
Not looking at embryos The Z4 Zoom stereoscope can also be used as a dissecting stereoscope when mounted on the standard dual-light stand. The upper and lower LED bulbs provide variable, cool, daylight-colored illumination for viewing true-color specimens and high-contrast structures. Popular in biology classrooms. Z4 stereo microscope, trinocular, dual LED stand, 110V
Catalog No. 50-005-84917 Supplier Med Vet International Supplier No. JOR755Z4T
Not looking at embryos The Z4 Zoom stereoscope can also be used as a dissecting stereoscope when mounted on the standard dual-light stand. The upper and lower LED bulbs provide variable, cool, daylight-colored illumination for viewing true-color specimens and high-contrast structures. Popular in biology classrooms. Z4 stereo microscope, trinocular, dual LED stand, 110V