Learn More
Medchemexpress LLC HY-126074 5mg Medchemexpress, Z433927330 CAS:1005883-72-6 Purity:>98%
SDP
Catalog No. 501932610
Encompass_Preferred
Medchemexpress, HY-126074 5mg Z433927330 CAS:1005883-72-6 Z433927330 is a potent and selective inhibitor of Aquaporin-7 (AQP7) , less potently inhibits AQP3 and AQPs9 , with IC 50 s of ~0.2 µM, ~0.7 µM and ~1.1 µM for mAQP7, mAQP3 and mAQP9, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2610 Supplier Medchemexpress LLC Supplier No. HY1260745MG
Medchemexpress, HY-126074 5mg Z433927330 CAS:1005883-72-6 Z433927330 is a potent and selective inhibitor of Aquaporin-7 (AQP7) , less potently inhibits AQP3 and AQPs9 , with IC 50 s of ~0.2 µM, ~0.7 µM and ~1.1 µM for mAQP7, mAQP3 and mAQP9, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.