Learn More
Selleck Chemical LLC Z57346765 hydrochloride E8275-1G
SDP
Catalog No. 5000042980
Encompass_Preferred
Z57346765 hydrochloride is an inhibitor of phosphoglycerate kinase 1 (PGK1) that binds to the ADP-binding pocket with a KD value of 2 09 10 M reducing PGK1 metabolic enzyme activity glucose consumption and lactate production It also inhibits DNA replication impairs DNA damage repair and induces cell cycle arrest in kidney renal clear cell carcinoma (KIRC) cells
Catalog No. 50-000-42980 Supplier Selleck Chemical LLC Supplier No. E82751G
Z57346765 hydrochloride is an inhibitor of phosphoglycerate kinase 1 (PGK1) that binds to the ADP-binding pocket with a KD value of 2 09 10 M reducing PGK1 metabolic enzyme activity glucose consumption and lactate production It also inhibits DNA replication impairs DNA damage repair and induces cell cycle arrest in kidney renal clear cell carcinoma (KIRC) cells