Learn More
Aviva Systems Biology Corp ZA20D3 Antibody - middle region (ARP34464_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34464_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZA20D3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZA20D3. Alternate names: AWP1, ZA20D3, ZFAND5BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52959