Learn More
Cayman Chemical ZA3-Ep10, Purity: ≥95%, CAS: 2090299-48-0, 1 mg
SDP
Catalog No. NC3714503
Encompass_Preferred
A zwitterionic aminolipid; has been used in the generation of LNPs for the delivery of mRNA and sgRNA in vitro and in vivo; LNPs containing ZA3-Ep10 selectively accumulate in the liver, lung, and kidneys over the heart and spleen in mice; ZA3-Ep10-containing LNPs encapsulating sgRNA targeting DNA encoding luciferase eliminate luciferase activity in HeLa-Luc-Cas9 cells
Catalog No. NC3714503 Supplier Cayman Chemical Supplier No. 407641 MG
A zwitterionic aminolipid; has been used in the generation of LNPs for the delivery of mRNA and sgRNA in vitro and in vivo; LNPs containing ZA3-Ep10 selectively accumulate in the liver, lung, and kidneys over the heart and spleen in mice; ZA3-Ep10-containing LNPs encapsulating sgRNA targeting DNA encoding luciferase eliminate luciferase activity in HeLa-Luc-Cas9 cells