Learn More
Selleck Chemical LLC Zabedosertib E2837-5MG
SDP
Catalog No. 5000040569
Encompass_Preferred
Zabedosertib (BAY 1834845) is a IRAK4 inhibitor with immunomodulatory potential which is a protein kinase involved in signaling innate immune responses from Toll-like receptors
Catalog No. 50-000-40569 Supplier Selleck Chemical LLC Supplier No. E28375MG
Zabedosertib (BAY 1834845) is a IRAK4 inhibitor with immunomodulatory potential which is a protein kinase involved in signaling innate immune responses from Toll-like receptors