Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zacopride hydrochloride | 101303-98-4 | MFCD04971977 | 99.7% | 346.25 | C15H21Cl2N3O2 | 1 ML
SDP

Catalog No. 5000246707
Encompass_Preferred

Zacopride hydrochloride is a research-grade 5-HT3 receptor antagonist supplied as either a 10 mM solution in DMSO or as solid material for pharmacological and analytical studies. It has defined purity and physicochemical properties suitable for in vitro and in vivo protocols. Intended for research use only.

  • High purity (99.69%).
  • Concentration: 10 mM solution, 1 mL vial in DMSO.
  • Also available as solid in multiple masses (5-100 mg; larger sizes by quote).
  • Molecular weight 346.25; formula C15H21Cl2N3O2.
  • CAS number 101303-98-4.
  • Recommended solvent: DMSO; in vivo formulation suggestions provided.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-46707 Supplier Medchemexpress LLC Supplier No. HY10313710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zacopride hydrochloride is a research-grade 5-HT3 receptor antagonist supplied as either a 10 mM solution in DMSO or as solid material for pharmacological and analytical studies. It has defined purity and physicochemical properties suitable for in vitro and in vivo protocols. Intended for research use only.

  • High purity (99.69%).
  • Concentration: 10 mM solution, 1 mL vial in DMSO.
  • Also available as solid in multiple masses (5-100 mg; larger sizes by quote).
  • Molecular weight 346.25; formula C15H21Cl2N3O2.
  • CAS number 101303-98-4.
  • Recommended solvent: DMSO; in vivo formulation suggestions provided.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.