Learn More
Medchemexpress LLC Zacopride hydrochloride | 101303-98-4 | MFCD04971977 | 99.7% | 346.25 g/mol | C15H21Cl2N3O2 | 10 MG
SDP
Catalog No. 5000246140
Encompass_Preferred
Zacopride hydrochloride is the hydrochloride salt of zacopride, a research reagent that functions as a 5-HT3 receptor antagonist and 5-HT4 receptor ligand. It is provided as a high-purity solid for use in receptor pharmacology and related in vitro and in vivo studies.
  • High purity suitable for analytical and pharmacological research.
  • Available in small solid quantities for assay development and dosing.
  • Well-characterized chemical identity and molecular weight for analytical workflows.
  • Appropriate for receptor binding, functional assays, and profiling studies.
  • Solid form simplifies storage and handling for laboratory use.
Catalog No. 50-002-46140 Supplier Medchemexpress LLC Supplier No. HY10313710MG
Zacopride hydrochloride is the hydrochloride salt of zacopride, a research reagent that functions as a 5-HT3 receptor antagonist and 5-HT4 receptor ligand. It is provided as a high-purity solid for use in receptor pharmacology and related in vitro and in vivo studies.
  • High purity suitable for analytical and pharmacological research.
  • Available in small solid quantities for assay development and dosing.
  • Well-characterized chemical identity and molecular weight for analytical workflows.
  • Appropriate for receptor binding, functional assays, and profiling studies.
  • Solid form simplifies storage and handling for laboratory use.