Learn More
Medchemexpress LLC Zacopride hydrochloride | 101303-98-4 | MFCD04971977 | 99.7% | 346.25 g/mol | C15H21Cl2N3O2 | 25 MG
SDP
Catalog No. 5000247609
Encompass_Preferred
Zacopride hydrochloride is a research-grade small molecule acting as a potent 5-HT3 receptor antagonist and a 5-HT4 receptor agonist, supplied for preclinical and in vitro studies. It is provided at research-grade purity and in small pack sizes suitable for laboratory use.
  • Orally active 5-HT3 receptor antagonist and 5-HT4 receptor agonist.
  • High purity (99.69%).
  • Available in small research pack sizes, including 25 MG.
  • Molecular weight 346.25 g/mol; formula C15H21Cl2N3O2.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-47609 Supplier Medchemexpress LLC Supplier No. HY10313725MG
Zacopride hydrochloride is a research-grade small molecule acting as a potent 5-HT3 receptor antagonist and a 5-HT4 receptor agonist, supplied for preclinical and in vitro studies. It is provided at research-grade purity and in small pack sizes suitable for laboratory use.
  • Orally active 5-HT3 receptor antagonist and 5-HT4 receptor agonist.
  • High purity (99.69%).
  • Available in small research pack sizes, including 25 MG.
  • Molecular weight 346.25 g/mol; formula C15H21Cl2N3O2.
  • Intended for research use only; not for human or veterinary use.