Learn More
Medchemexpress LLC Zacopride hydrochloride | 101303-98-4 | MFCD20488057 | 99.7% | 346.25 | C15H21Cl2N3O2 | 50 MG
SDP
Catalog No. 5000247024
Encompass_Preferred
Zacopride hydrochloride (CAS 101303-98-4) is a research-grade serotonergic agent used primarily as a 5-HT3 receptor antagonist and 5-HT4 receptor agonist. It is supplied as an analytical standard for laboratory research in gastrointestinal, cardiovascular, and neuropsychiatric studies.
  • High purity suitable for analytical applications.
  • Well-characterized pharmacological profile as a 5-HT3 antagonist and 5-HT4 agonist.
  • Commonly used in studies of gastrointestinal motility and cardiac function.
  • Provided in small research pack sizes for bench-scale experiments.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-47024 Supplier Medchemexpress LLC Supplier No. HY10313750MG
Zacopride hydrochloride (CAS 101303-98-4) is a research-grade serotonergic agent used primarily as a 5-HT3 receptor antagonist and 5-HT4 receptor agonist. It is supplied as an analytical standard for laboratory research in gastrointestinal, cardiovascular, and neuropsychiatric studies.
  • High purity suitable for analytical applications.
  • Well-characterized pharmacological profile as a 5-HT3 antagonist and 5-HT4 agonist.
  • Commonly used in studies of gastrointestinal motility and cardiac function.
  • Provided in small research pack sizes for bench-scale experiments.
  • Intended for research use only; not for human or clinical use.