Learn More
Medchemexpress LLC Zacopride hydrochloride | 101303-98-4 | MFCD20488057 | 99.7% | 346.3 g/mol | C15H20ClN3O2 · HCl | 100 MG
SDP
Catalog No. 5000247205
Encompass_Preferred
Zacopride hydrochloride is the hydrochloride salt of zacopride, a research compound that functions as a potent 5-HT3 receptor antagonist and a 5-HT4 receptor agonist. It is supplied as a white to off-white solid for laboratory research use in studies of gastrointestinal, cardiovascular, and neuropsychiatric biology.
  • Research reagent for receptor pharmacology and in vitro assays.
  • High purity suitable for analytical and preclinical studies.
  • Solid form, soluble in dimethyl sulfoxide (DMSO) for stock solution preparation.
  • Packaged in a small 100 mg quantity for bench-scale experiments.
Catalog No. 50-002-47205 Supplier Medchemexpress LLC Supplier No. HY103137100MG
Add to Cart
Add to Cart
Zacopride hydrochloride is the hydrochloride salt of zacopride, a research compound that functions as a potent 5-HT3 receptor antagonist and a 5-HT4 receptor agonist. It is supplied as a white to off-white solid for laboratory research use in studies of gastrointestinal, cardiovascular, and neuropsychiatric biology.
  • Research reagent for receptor pharmacology and in vitro assays.
  • High purity suitable for analytical and preclinical studies.
  • Solid form, soluble in dimethyl sulfoxide (DMSO) for stock solution preparation.
  • Packaged in a small 100 mg quantity for bench-scale experiments.