Learn More
TARGETMOL CHEMICALS INC ZACOPRIDE HYDROCHLORIDE 10MG
SDP
Catalog No. 502579787
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zacopride is a highly potent 5-HT3 receptor antagonist (Kd = 0.38 nM) and 5-HT4 receptor agonist (Ki = 373 nM). It also is a selective IK1 channel agonist. purity: 98%
Catalog No. 50-257-9787 Supplier TARGETMOL CHEMICALS INC Supplier No. T533610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zacopride is a highly potent 5-HT3 receptor antagonist (Kd = 0.38 nM) and 5-HT4 receptor agonist (Ki = 373 nM). It also is a selective IK1 channel agonist. purity: 98%