Learn More
Apexbio Technology LLC Zacopride hydrochloride 101303-98-4 10mg
SDP
Catalog No. 502240188
Encompass_Preferred
Zacopride hydrochloride (CAS 101303-98-4) is a small molecule that functions as a high-affinity antagonist at 5-HT sub 3 /sub receptors ( i K sub i /sub /i 0 38 nM) and as an agonist at 5-HT sub 4 /sub receptors ( i K sub i /sub /i 373 nM) These receptors are subtypes of serotonin (5-HT) receptors involved in modulating neurotransmission in central and peripheral systems In human adrenocortical tissue zacopride hydrochloride induces aldosterone secretion in a dose-dependent manner with an effective concentration as low as 10 sup -10 /sup mol/L and half-maximal stimulation at 7 10 sup -8 /sup mol/L In vivo oral administration of 400 g zacopride increased plasma aldosterone without affecting renin adrenocorticotropic hormone or cortisol Zacopride hydrochloride is valuable for studying serotonergic regulation of adrenal steroidogenesis and receptor pharmacology
This item is not returnable. View return policy
Catalog No. 50-224-0188 Supplier Apexbio Technology LLC Supplier No. B685810
Zacopride hydrochloride (CAS 101303-98-4) is a small molecule that functions as a high-affinity antagonist at 5-HT sub 3 /sub receptors ( i K sub i /sub /i 0 38 nM) and as an agonist at 5-HT sub 4 /sub receptors ( i K sub i /sub /i 373 nM) These receptors are subtypes of serotonin (5-HT) receptors involved in modulating neurotransmission in central and peripheral systems In human adrenocortical tissue zacopride hydrochloride induces aldosterone secretion in a dose-dependent manner with an effective concentration as low as 10 sup -10 /sup mol/L and half-maximal stimulation at 7 10 sup -8 /sup mol/L In vivo oral administration of 400 g zacopride increased plasma aldosterone without affecting renin adrenocorticotropic hormone or cortisol Zacopride hydrochloride is valuable for studying serotonergic regulation of adrenal steroidogenesis and receptor pharmacology