Learn More
Aviva Systems Biology Corp ZADH1 Antibody - middle region (ARP52975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZADH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZADH1. Alternate names: DKFZp686P10120, FLJ39091, PGR-2, PGR2, ZADH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60885