Learn More
Apexbio Technology LLC Zafirlukast 107753-78-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20685.1
Encompass_Preferred
Zafirlukast (CAS 107753-78-6) is an antagonist targeting cysteinyl leukotriene receptors (CysLTR) which mediate inflammatory signaling pathways By competitively inhibiting leukotriene receptor binding it disrupts the leukotriene-driven inflammatory response Zafirlukast demonstrates antagonist potency against leukotriene receptors with an IC50 value of approximately 0 6 M and exhibits moderate inhibitory activity on CYP2C9 enzyme (IC50 of about 7 0 M) In biomedical research Zafirlukast serves as a pharmacological tool for studying leukotriene-mediated inflammation and receptor signaling processes
Catalog No. 50-000-62598