Learn More
Agilent Technologies ZAG 130 dsDNA Separation Gel, 500 mL
Catalog No. NC1790311
Encompass
These separation gels are analytical reagents designed for high-resolution DNA fragment analysis. Engineered for consistent performance, these liquid-state gels support applications including PCR fragment sizing, genotyping, and microsatellite detection. They provide precise sizing across a broad range of DNA fragments and are optimized for rapid separation protocols.
  • Broad sizing range from 1 to 20,000 bp
  • High resolution up to 3 bp
  • Fast separation times of 30 minutes
  • Qualitative range from 0.5 to 50 ng/μL
  • Ready-to-use liquid format
Catalog No. NC1790311 Supplier Agilent Technologies Supplier No. ZAG1300500
These separation gels are analytical reagents designed for high-resolution DNA fragment analysis. Engineered for consistent performance, these liquid-state gels support applications including PCR fragment sizing, genotyping, and microsatellite detection. They provide precise sizing across a broad range of DNA fragments and are optimized for rapid separation protocols.
  • Broad sizing range from 1 to 20,000 bp
  • High resolution up to 3 bp
  • Fast separation times of 30 minutes
  • Qualitative range from 0.5 to 50 ng/μL
  • Ready-to-use liquid format