Learn More
Agilent Technologies ZAG 110 dsDNA Kit (35-5000bp)
Supplier:  Agilent Technologies ZAG1105000
Encompass
ZAG 110 dsDNA Kit (35-5000bp), 5000. ZAG DNA Analyzer system kit for genotyping and qualitative analysis of PCR amplicons from 35 to 5,000 bp. Includes reagents necessary for running 5,000 samples.
Catalog No. NC1788278