Learn More
AdipoGen Zinc-alpha-2-glycoprotein (human) Matched Pair Detection Set (1 Set)
SDP
Catalog No. 502053339
Encompass_Preferred
Matched Pair Detection Set. Detects human Zinc-alpha-2-glycoprotein. Does not detect mouse Zinc-alpha-2-glycoprotein. Colorimetric assay. Sample Type: Cell Culture Supernatant, Plasma, Serum. Range: 0.0156 to 1 ng/ml. Sensitivity: 100 pg/ml. ZAG stimulates lipolysis in in vitro and in vivo experiments. Due to its expression in, and secretion from adipocytes, ZAG is considered an adipokine. Recently, the clinical significance of ZAG has been clarified. ZAG expression in adipocytes is inversely related to fat mass, thus it is intimately involved in the maintenance of body weight in mice and humans. Epidemiological studies have uncovered an association between ZAG and plasma cholesterol. The non-synonymous single nucleotide polymorphism rs4215 in ZAG is associated with plasma cholesterol and obesity.
Catalog No. 50-205-3339 Supplier AdipoGen Supplier No. AG46B0008KI01
Matched Pair Detection Set. Detects human Zinc-alpha-2-glycoprotein. Does not detect mouse Zinc-alpha-2-glycoprotein. Colorimetric assay. Sample Type: Cell Culture Supernatant, Plasma, Serum. Range: 0.0156 to 1 ng/ml. Sensitivity: 100 pg/ml. ZAG stimulates lipolysis in in vitro and in vivo experiments. Due to its expression in, and secretion from adipocytes, ZAG is considered an adipokine. Recently, the clinical significance of ZAG has been clarified. ZAG expression in adipocytes is inversely related to fat mass, thus it is intimately involved in the maintenance of body weight in mice and humans. Epidemiological studies have uncovered an association between ZAG and plasma cholesterol. The non-synonymous single nucleotide polymorphism rs4215 in ZAG is associated with plasma cholesterol and obesity.