Learn More
AdipoGen Zinc-alpha-2-glycoprotein (human) (rec.) (untagged) (3 x 50 µg)
SDP
Supplier:  AdipoGen AG40B01463050
Encompass_Preferred
Protein. Human full-length zinc-alpha-2-glycoprotein (aa 21-298) is untagged. Source: E. coli. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). ZAG is considered an adipokine. ZAG expression in adipocytes is inversely related to fat mass, thus it is intimately involved in the maintenance of body weight in mice and humans. Epidemiological studies have uncovered an association between ZAG and plasma cholesterol. The non-synonymous single nucleotide polymorphism rs4215 in ZAG is associated with plasma cholesterol and obesity. Structurally ZAG possesses a class I major histocompatibility complex (MHC) protein fold. It is distinct from other members of this protein family in that it is soluble, rather than being anchored to plasma membranes, and it associates with prolactin inducible protein rather than beta2-microglobulin.
Catalog No. 50-205-3233