Learn More
Medchemexpress LLC Zagociguat (CY-6463) | 2201048-82-8 | MFCD34676490 | 98.0% | 362.28 g/mol | C16H10F4N6 | 5 MG
SDP
Catalog No. 5000221015
Encompass_Preferred
Zagociguat is an orally active, blood-brain barrier-permeable soluble guanylate cyclase (sGC) stimulator used in research to elevate intracellular cGMP and investigate NO-sGC-cGMP signaling pathways. It is employed in preclinical and clinical research addressing neurodegeneration and mitochondrial disease and is supplied in small research pack sizes with defined storage conditions.
  • Orally active and brain-penetrant sGC stimulator.
  • Increases intracellular cGMP to modulate NO-sGC-cGMP signaling.
  • Applicable to neurodegeneration and mitochondrial disease research.
  • Provided in small research pack sizes suitable for laboratory studies.
  • Powder stable at -20°C for up to 3 years; solutions require low-temperature storage.
Catalog No. 50-002-21015 Supplier Medchemexpress LLC Supplier No. HY1456075MG
Zagociguat is an orally active, blood-brain barrier-permeable soluble guanylate cyclase (sGC) stimulator used in research to elevate intracellular cGMP and investigate NO-sGC-cGMP signaling pathways. It is employed in preclinical and clinical research addressing neurodegeneration and mitochondrial disease and is supplied in small research pack sizes with defined storage conditions.
  • Orally active and brain-penetrant sGC stimulator.
  • Increases intracellular cGMP to modulate NO-sGC-cGMP signaling.
  • Applicable to neurodegeneration and mitochondrial disease research.
  • Provided in small research pack sizes suitable for laboratory studies.
  • Powder stable at -20°C for up to 3 years; solutions require low-temperature storage.