Learn More
Cayman Chemical ZalcItabIn 100mg
SDP
Catalog No. 502971297
Encompass_Preferred
A NRTI; reduces the replication of laboratory-adapted and clinical isolates of HIV in a plaque reduction assay (IC50s = 0.013-0.08 µM)
Catalog No. 50-297-1297 Supplier Cayman Chemical Supplier No. 16019100 MG
A NRTI; reduces the replication of laboratory-adapted and clinical isolates of HIV in a plaque reduction assay (IC50s = 0.013-0.08 µM)