Learn More
Apexbio Technology LLC Zalcitabine 7481-89-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062084
Encompass_Preferred
Zalcitabine is a nucleoside analog inhibitor targeting viral reverse transcriptase a key enzyme promoting viral replication By competitively binding to the active site of reverse transcriptase zalcitabine terminates DNA chain elongation effectively inhibiting viral DNA synthesis In vitro studies have demonstrated inhibitory activity against HIV-1 with reported IC50 values ranging approximately from 0 03 to 0 3 M Zalcitabine is frequently utilized in biomedical research as an antiviral reference molecule for studying HIV therapeutic targets antiviral drug resistance and mechanisms of reverse transcription inhibition
Catalog No. 50-000-62084 Supplier Apexbio Technology LLC Supplier No. B22235.1
Zalcitabine is a nucleoside analog inhibitor targeting viral reverse transcriptase a key enzyme promoting viral replication By competitively binding to the active site of reverse transcriptase zalcitabine terminates DNA chain elongation effectively inhibiting viral DNA synthesis In vitro studies have demonstrated inhibitory activity against HIV-1 with reported IC50 values ranging approximately from 0 03 to 0 3 M Zalcitabine is frequently utilized in biomedical research as an antiviral reference molecule for studying HIV therapeutic targets antiviral drug resistance and mechanisms of reverse transcription inhibition