Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zaldaride maleate | 109826-27-9 | 99.7% | 544.60 g/mol | C30H32N4O6 | 1 ML
SDP

Catalog No. 5000243884
Encompass_Preferred

Zaldaride maleate is a potent, orally active, and selective calmodulin inhibitor provided as a solid and as a ready-to-use 10 mM solution (1 mL). It is supplied with high reported purity and a documented IC50 in the low nanomolar range, and is intended for biochemical and pharmacological research involving calmodulin-mediated pathways. Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

  • Potent calmodulin inhibitor with low-nanomolar activity.
  • Offered as a solid and as a 10 mM solution (1 mL) for convenience.
  • High reported purity suitable for research use.
  • Molecular weight approximately 544.60 g/mol.
  • Storage guidelines for sealed material and solutions provided.

Catalog No. 50-002-43884 Supplier Medchemexpress LLC Supplier No. HY105118A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zaldaride maleate is a potent, orally active, and selective calmodulin inhibitor provided as a solid and as a ready-to-use 10 mM solution (1 mL). It is supplied with high reported purity and a documented IC50 in the low nanomolar range, and is intended for biochemical and pharmacological research involving calmodulin-mediated pathways. Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

  • Potent calmodulin inhibitor with low-nanomolar activity.
  • Offered as a solid and as a 10 mM solution (1 mL) for convenience.
  • High reported purity suitable for research use.
  • Molecular weight approximately 544.60 g/mol.
  • Storage guidelines for sealed material and solutions provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.